Learn More
Electron Microscopy Sciences Phosphate Buffer, pH 6.5 500 ML
SDP
Catalog No. 5031700
Encompass_Preferred
Used in Differentiation of leucocytes, rickettsia, bacteria and inclusion bodies. Nuclei, additional leukocyte features are similar. References: Clark, G., Staining Procedures, Williams & Wilkins Co., Baltimore, 3rd Ed., p. 125, c. 1973
Catalog No. 50-317-00 Supplier Electron Microscopy Sciences Supplier No. 2615302
Used in Differentiation of leucocytes, rickettsia, bacteria and inclusion bodies. Nuclei, additional leukocyte features are similar. References: Clark, G., Staining Procedures, Williams & Wilkins Co., Baltimore, 3rd Ed., p. 125, c. 1973