Learn More
MOLTOX Molecular Toxicology Inc Phosphate Buffer - 0.2M Sodium Phosphate, pH 7.4, 500 mL/bottle, Store at Room Temperature
SDP
Catalog No. 50948735
Encompass
Butterfield""s Phosphate buffer (pH 7.2) is a standard medium used for the preparation of standard dilutions in laboratory procedures. This product is also used in water and microbial limit testing. MOLTOX offers a wide array of solutions and buffers for a variety of laboratory research needs. All reagents are accompanied by a GLP-compliant Formulation and Quality Control statement.
Catalog No. 50-948-735 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 26510.047
Butterfield""s Phosphate buffer (pH 7.2) is a standard medium used for the preparation of standard dilutions in laboratory procedures. This product is also used in water and microbial limit testing. MOLTOX offers a wide array of solutions and buffers for a variety of laboratory research needs. All reagents are accompanied by a GLP-compliant Formulation and Quality Control statement.