Learn More
MOLTOX Molecular Toxicology Inc Phosphate Buffer (Butterfields), pH 7.2, 1000 mL/bottle, Store at Room Temperature
SDP
Catalog No. 50948785
Encompass_Preferred
Butterfield""s Phosphate buffer (pH 7.2) is a standard medium used for the preparation of standard dilutions in laboratory procedures. This product is also used in water and microbial limit testing. MOLTOX manufactures a wide variety of buffers for your laboratory needs. All MOLTOX reagents are accompanied by a GLP-compliant Formulation and Quality Control statement.
Catalog No. 50-948-785 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 26622
Butterfield""s Phosphate buffer (pH 7.2) is a standard medium used for the preparation of standard dilutions in laboratory procedures. This product is also used in water and microbial limit testing. MOLTOX manufactures a wide variety of buffers for your laboratory needs. All MOLTOX reagents are accompanied by a GLP-compliant Formulation and Quality Control statement.