Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphate Buffer, 0.1 M, pH 2.5, Ricca Chemical
SDP

Catalog No. R57881001A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R57881001A 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R57881001A Supplier Ricca Chemical Company Supplier No. R57881001A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A general laboratory reagent for pH control applications.

Chemical Identifiers

CAS 7778-77-0
Molecular Formula H2KO4P
Molecular Weight (g/mol) 136.08
MDL Number MFCD00011401 MFCD00147253
InChI Key GNSKLFRGEWLPPA-UHFFFAOYSA-M
PubChem CID 516951
ChEBI CHEBI:63036
IUPAC Name potassium dihydrogen phosphate
SMILES [K+].OP(O)([O-])=O

Specifications

Color Clear colorless liquid
Concentration 0.1 M
For Use With (Application) Water Analysis
pH 2.5
Physical Form Liquid
Quantity 1 L
Solution Type Buffer
Odor Odorless
Packaging Natural Poly Bottle
Solubility Information Solubility in Water: Infinite
Grade Laboratory
Show More Show Less
Recommended Storage General
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.