Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphate Buffer Solution, pH 7, Solstice

Catalog No. 60048561 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Poly bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-048-561 1 L Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-048-561 Supplier Solstice Supplier No. 731731L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7732-18-5

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7732-18-5

Specifications

Density 1.01g/mL at 20°C
For Use With (Application) pH Calibration
pH 6.98 to 7.02
Physical Form Liquid
Quantity 1 L
Solution Type pH Buffer
Packaging Poly bottle
UN Number NONH for all modes of transport
Grade Not Available
Chemical Name or Material Phosphate buffer solution
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.