Learn More
VWR INTERNATIONAL LLC PHOSPHATE BUFFERD SALINE 500ML
Catalog No. NC9630775
Encompass
PHOSHPHATE BUFFERED SALINE (PBS), STERILE LIQUID. 500ml. 1X Sterile PBS, pH 7.4. 137 mM sodium chloride, 2.7 mM potassium chloride, 12 mM phosphate buffer.
Catalog No. NC9630775 Supplier VWR INTERNATIONAL LLC Supplier No. 97062818
PHOSHPHATE BUFFERED SALINE (PBS), STERILE LIQUID. 500ml. 1X Sterile PBS, pH 7.4. 137 mM sodium chloride, 2.7 mM potassium chloride, 12 mM phosphate buffer.