Learn More
Fd Neurotechnologies Inc PHOSPHATE-BUFFERED 4% PARAFORM
SDP
Catalog No. NC0590520
Encompass
This 1000ml filtered and ready-to-use tissue fixative is freshly made from high purity paraformaldehyde powder. This solution may be stored at room temperature for at least one year without change of the original quality. The standard solution contains 4% paraformaldehyde in 0.1 M phosphate buffer (pH 7.4). Other concentrations can also be provided upon request.
Catalog No. NC0590520 Supplier Fd Neurotechnologies Inc Supplier No. PF101
This 1000ml filtered and ready-to-use tissue fixative is freshly made from high purity paraformaldehyde powder. This solution may be stored at room temperature for at least one year without change of the original quality. The standard solution contains 4% paraformaldehyde in 0.1 M phosphate buffer (pH 7.4). Other concentrations can also be provided upon request.