Learn More
AG Scientific Inc Phosphate Buffered Saline 1 L
Catalog No. 502268353
Encompass_Preferred
Phosphate Buffered Saline is a buffer solution commonly used in biological research. PBS has many uses because it is isotonic and non-toxic to most cells. These uses include substance dilution and cell container rinsing.Ready to use sterile 1X solution, pH: 7.25 - 7.55
Catalog No. 50-226-8353 Supplier AG Scientific Inc Supplier No. P24261L
Phosphate Buffered Saline is a buffer solution commonly used in biological research. PBS has many uses because it is isotonic and non-toxic to most cells. These uses include substance dilution and cell container rinsing.Ready to use sterile 1X solution, pH: 7.25 - 7.55