Learn More
Sigma Aldrich Fine Chemicals Biosciences Phosphate buffered saline
Catalog No. 502900914
Encompass
Phosphate buffered saline (PBS) is an isotonic buffer SOLUTIONon. This water-based salt SOLUTIONon is made up of disodium hydrogen phosphate sodium chloride and in some preparations potassium chloride and potassium dihydrogen phosphate. PBS is generally used in biological research.
Catalog No. 50-290-0914 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P5119100ML
Phosphate buffered saline (PBS) is an isotonic buffer SOLUTIONon. This water-based salt SOLUTIONon is made up of disodium hydrogen phosphate sodium chloride and in some preparations potassium chloride and potassium dihydrogen phosphate. PBS is generally used in biological research.