Learn More
Sigma Aldrich Fine Chemicals Biosciences Phosphate buffered saline, 10-pack
Catalog No. NC9865688
Encompass
Phosphate buffered saline, powder, pH 7.4, for preparing 1 L solutions, 10-pack
Catalog No. NC9865688 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P381310PAK
Phosphate buffered saline, powder, pH 7.4, for preparing 1 L solutions, 10-pack