Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphate Buffered Saline, 1X - Non-sterile, Ricca Chemical
SDP

Catalog No. R58190001A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R58190001A 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R58190001A Supplier Ricca Chemical Company Supplier No. R58190001A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Phosphate Buffered Saline, 1X is non-sterile.

  • Non-sterile saline

Specifications

Chemical Name or Material Phosphate Buffered Saline (PBS)
Melting Point ∼ 0°C
Color Colorless
Concentration 1X
For Use With (Application) in Vitro Diagnostics
pH 7.35 to 7.45
Physical Form Liquid
Quantity 1 L
Solution Type Phosphate Buffered Saline
Packaging Natural Poly Bottle
Sterility Non-sterile
Grade Chemical Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.