Learn More
Medchemexpress LLC Phosphate-Buffered Saline | 500 ML
SDP
Catalog No. 5000428945
Encompass_Preferred
Phosphate-Buffered Saline (PBS) is a balanced salt solution designed for various cell culture applications. These applications include washing cells, diluting cells, and preparing reagents. The standard base specification for this product is 500 mL, with larger sizes being incremental volumes of this base.
  • Filtered and ready to use for general cell culture purposes.
  • Free of Mg2+, Ca2+, and phenol red.
  • pH is 7.2-7.4.
Catalog No. 50-004-28945 Supplier Medchemexpress LLC Supplier No. HYK3005500ML 6
Phosphate-Buffered Saline (PBS) is a balanced salt solution designed for various cell culture applications. These applications include washing cells, diluting cells, and preparing reagents. The standard base specification for this product is 500 mL, with larger sizes being incremental volumes of this base.
  • Filtered and ready to use for general cell culture purposes.
  • Free of Mg2+, Ca2+, and phenol red.
  • pH is 7.2-7.4.