Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Phosphate-buffered saline (DPBS, 10X), Dulbecco's formula

Catalog No. p-7076577
Change view
Click to view available options
Quantity:
500 mL
1 L
2 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61917AP 500 mL
AAJ61917K2 1 L
AAJ61917K3 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ61917AP Supplier Thermo Scientific Chemicals Supplier No. J61917AP
Only null left
Add to Cart

Used as a buffer system to maintain cell culture media in the physiological pH range. In combination with glucose, it provides the principal energy sources of cell metabolism. It is involved to irrigate, wash and dilute mammalian cells.

Phosphate-buffered saline (DPBS, 10X), Dulbecco's formula is used as a buffer system to maintain cell culture media in the physiological pH range. In combination with glucose, it provides the principal energy sources of cell metabolism. It is involved to irrigate, wash and dilute mammalian cells. It plays an important role in the transportation of tissue samples and as diluents in various reagent preparations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Quantity 500 mL
Packaging Plastic bottle
What are the concentrations of the components in Phosphate-buffered saline (DPBS, 10X), Dulbecco's formula?

Phosphate-buffered saline (DPBS, 10X), Dulbecco's formula contains 89 mM sodium phosphate dibasic, 15 mM potassium phosphate monobasic, 1.37 mM sodium chloride, and 27 mM potassium chloride.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.