Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Phosphate-buffered saline (DPBS, 1X), Dulbecco's formula, without calcium, without magnesium

Catalog No. p-7076758
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ67802K2 1 L
AAJ67802AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ67802K2 Supplier Thermo Scientific Chemicals Supplier No. J67802K2
Add to Cart

missing translation for 'validMainframeMsg'

An anticoagulant and blood preservative. Permits the storage of whole blood at refrigerator temperatures for approximately 10 weeks.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Physical Form Liquid
Quantity 1 L
Packaging Plastic bottle

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.