Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Phosphate-buffered saline (PBS, 10X), pH 7.4

Catalog No. AAJ62036K3
Change view
Click to view available options
Quantity:
1 L
2 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ62036K3 2 L
AAJ62036K2 1 L
AAJ62036K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ62036K3 Supplier Thermo Scientific Chemicals Supplier No. J62036K3
Add to Cart

missing translation for 'validMainframeMsg'

Non-toxic to most cells and is used in biological research.

Phosphate-buffered saline (PBS, 10X) is an isotonic, non-toxic to most cells and is used in biological research. It is associated with ethylenediaminetetraacetic acid (EDTA) and is used to disengage attached and clumped cells. It also helps to maintain a constant pH.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phosphate Buffered Saline (PBS)
Color Colorless
Concentration 10 X
pH 7.4
Physical Form Liquid
Quantity 2 L
What is the composition of Phosphate-buffered saline (PBS, 10X), pH 7.4 (Cat. No. J62036.K7, J62036-K7, J62036.K2, J62036-K2, J62036.K3, J62036-K3)?

Here is the composition of Phosphate-buffered saline (PBS, 10X), pH 7.4 (Cat. No. J62036.K7, J62036-K7, J62036.K2, J62036-K2, J62036.K3, J62036-K3):

- Sodium chloride: 1.37 M
- Potassium chloride (KCl): 27 mM
- Phosphate buffer: 101.4 mM
- Potassium phosphate monobasic: 17.6 mM

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.