Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Phosphate-buffered saline (PBS, 5X), with EDTA

Catalog No. AAJ60893K3
Change view
Click to view available options
Quantity:
1 L
2 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60893K3 2 L
AAJ60893K2 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60893K3 Supplier Thermo Scientific Chemicals Supplier No. J60893K3
  • $149.60
    $127.65 / Each of 1

    Save $21.95

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

Helps to maintain a constant pH

PBS with EDTA is also used to disengage attached and clumped cells. PBS-EDTA is used as a buffer in procedures generating dendritic cells from monocytes. PBS+EDTA to get better cell sorting. PBS/EDTA may also be used directly without addition of trypsine for very gentle cell dissociation. PBS can be used to take a reference spectrum when measuring the protein adsorption in ellipsometry. PBS can be used as a diluent in methods to dry biomolecules.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Phosphate Buffered Saline (PBS)
Concentration 5 X
Physical Form Liquid
Quantity 2 L

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.