Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-Biosciences Phosphate Buffered Saline (PBS), Sterile, [1X]
SCI_ED

Catalog No. S02029
Change view
Click to view available options
Quantity:
10 L
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02029 10 L
S02028 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S02029 Supplier G-Biosciences Supplier No. 7861945
Add to Cart
Add to Cart

1X PBS (Phosphate Buffered Saline) is a ready-to-use, sterile, pH adjusted blend of phosphate buffer saline.

  • 1X PBS is a ready-to-use phosphate buffer saline solution of 8mM NaH2PO4, 150mM NaCl, 3mM KCl, and 2mM KH2PO4 at pH 7.4.
  • The 1X PBS solution is a sterile solution.

Specifications

Chemical Name or Material Buffer Solution
For Use With (Equipment) ArcturusXT LCM Instrument
Physical Form Liquid
Quantity 10 L
Solution Type Buffer
Sterility Sterile
Grade Chemical Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.