Learn More
Boston Bioproducts Inc Phosphate Buffered Saline, PBS (10X, pH 7.6) - 1L
Catalog No. NC0442092
Encompass
Phosphate Buffered Saline (PBS) is a buffer solution widely used in biological research due to its isotonic and non-toxic nature. With a physiological pH range, PBS mimics the osmolarity and ion concentrations of the human body, making it ideal for applications including protein purification, Western Blotting and immunoassays, sample dilution, and as a transport medium. This buffer is formulated in a 10X concentration, and can be diluted to support its application.
Catalog No. NC0442092 Supplier Boston Bioproducts Inc Supplier No. BM2211L
Phosphate Buffered Saline (PBS) is a buffer solution widely used in biological research due to its isotonic and non-toxic nature. With a physiological pH range, PBS mimics the osmolarity and ion concentrations of the human body, making it ideal for applications including protein purification, Western Blotting and immunoassays, sample dilution, and as a transport medium. This buffer is formulated in a 10X concentration, and can be diluted to support its application.