Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab Phosphate Buffered Saline, pH 7.2
SDP

Catalog No. 22046621
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-046-621 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Harmful if swallowed, in contact with skin, or if inhaled
Catalog No. 22-046-621 Supplier StatLab Supplier No. BUP0250
Add to Cart
Add to Cart
Harmful if swallowed, in contact with skin, or if inhaled

Buffers are made from the highest quality chemicals using a specialized process which ensures exact pH levels.

  • Use as a washing buffer during immunohistochemistry (IHC) and immunofluorescence to remove unbound antibodies without altering tissue structure.
  • Also use as a diluent for preparing fixatives like paraformaldehyde.
  • StatLab MasterTech special stain kit component.

Specifications

For Use With (Equipment) Electrode
Quantity 1 L
Solution Type Aqueous
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.