Learn More
Thermo Scientific™ Phosphate Buffered Saline with Poly 80 (PBST)
Catalog No. R112502
Change view
Click to view available options
Description:
Phosphate buffered saline with Poly 80 (PBST)
Quantity:
10/Pk.
Fill Volume:
50 mL
Catalog No. Description Quantity Fill Volume
R112502 Phosphate buffered saline with Poly 80 (PBST) 10/Pk. 50 mL
1 options
This item is not returnable. View return policy
Catalog No. R112502 Supplier Thermo Scientific™ Supplier No. R112502
Add to Cart
Add to Cart
This item is not returnable. View return policy
Create a pH-balanced environment, much like the human body, and neutralize disinfectants and cleaning agents to aid organism recovery.
Recover microorganisms from multiple sample types, including rinse samples from clinical instruments and environmental monitoring samples, by creating a pH-balanced environment with Thermo Scientific™ Phosphate Buffered Saline with Poly 80 (PBST), a buffer solution commonly used in biological research.
The osmolarity and ion concentrations of Phosphate Buffered Saline with Poly 80 (PBST) solution usually match those of the human body (isotonic), creating an environment ideal for maintaining cell viability and the recovery and revitalization of injured or damaged cells. When polysorbate 80 is added, it provides a neutralizing property for disinfectants or cleaning solutions to further allow the recovery of cells.
Not all products are available for sale in all territories. Please inquire.
TRUSTED_SUSTAINABILITY
Specifications
Description Phosphate buffered saline with Poly 80 (PBST)
Product Type Buffer Solution
Fill Volume 50 mL
Quantity 10/Pk.
Laboratory Use only