Learn More
Sigma Aldrich Fine Chemicals Biosciences Phosphate Buffered Saline1L
Catalog No. 502899885
Encompass
Phosphate Buffered Saline (PBS) is routinely used as a wash buffer in Western blot and immunoprecipitation (IP) procedures as well as during sample preparation of cell lysates for use in general molecular biology applications.
Catalog No. 50-289-9885 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. P70591L
Phosphate Buffered Saline (PBS) is routinely used as a wash buffer in Western blot and immunoprecipitation (IP) procedures as well as during sample preparation of cell lysates for use in general molecular biology applications.