Learn More
MP Biomedicals™ Phosphate Buffered Saline (PBS), Dulbecco's formula, without Magnesium and Calcium
Shop All MP Biomedicals, Inc ProductsDescription
Specifications
Specifications
| Color | White |
| For Use With (Application) | Used for the handling and culturing of mammalian cells |
| pH | 7.4 |
| Physical Form | Powder |
| Quantity | 1 L |
| Formulation | Inorganic Salts: Potassium Chloride [KCl]: 200.0000 mg/L; Potassium Phosphate Monobasic [KH2PO4]: 200.0000 mg/L; Sodium Chloride [NaCl]: 8000.0000 mg/L; Sodium Phosphate Dibasic [Na2HPO4]: 1150.0000 mg/L |
| Osmolality | 279 ±15 mm/kg |
| Grade | Not Available |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.