Learn More
Thermo Scientific Dionex Inorganic Anion and Oxyhalide Standards | | 1 EA
Catalog No. NC1709226 Shop All Thermo Scientific Products
Encompass
Increase your productivity and reduce standard preparation errors with Thermo Scientific Dionex Ion Standards for routine anion and oxyhalide analyses. Made with high purity reagent-grade chemicals and water. Each standard is NIST traceable and includes a Certificate of Analysis.
Catalog No. NC1709226 Supplier Thermo Scientific Supplier No. 303172
Add to Cart
Add to Cart
Increase your productivity and reduce standard preparation errors with Thermo Scientific Dionex Ion Standards for routine anion and oxyhalide analyses. Made with high purity reagent-grade chemicals and water. Each standard is NIST traceable and includes a Certificate of Analysis.