Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CHEMetrics™ by AquaPhoenix Phosphate Refill Kit
SDP

Catalog No. 13299642
Change view
Click to view available options
Quantity:
30/Box
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
13-299-642 30/Box
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 13-299-642 Supplier CHEMetrics™ by AquaPhoenix Supplier No. R8515
Add to Cart
Add to Cart
This item is not returnable. View return policy

Refill kit for phosphate visual test kits used for ortho phosphate analysis in water. Contains 30 self-filling ampoules packaged securely in trays and a box.

  • In an acidic solution, ortho-phosphate reacts with ammonium molybdate and ammonium vanadate to produce a yellow colored complex in direct proportion to the phosphate concentration.
  • Results are expressed in ppm (mg/L) phosphate as PO4. To convert results from ppm PO4 to ppm P, divide by 3.06

Specifications

pH Range 7
Volume (Metric) 0.5 mL
For Use With (Equipment) K-8515 or K-8530
No. of Tests 30
Quantity 30/Box
Type Phosphate Refill Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.