Learn More
ERA WATERS CORP PHOSPHATE STD 1000MG/L 500ML
SDP
Catalog No. NC9738453
Encompass
One 500 mL standard containing Phosphate as P at 1000 mg/L in H2O. This CRM is covered by ERA's ISO Guide 34 accreditation.
Catalog No. NC9738453 Supplier ERA WATERS CORP Supplier No. 993
One 500 mL standard containing Phosphate as P at 1000 mg/L in H2O. This CRM is covered by ERA's ISO Guide 34 accreditation.