Learn More
Hach Company Phosphate Standard Solution, 100 mg/L as PO4 (NIST), 100 mL MDB
Catalog No. 501995006 Shop All Hach Company Products
Encompass
For accuracy checks in phosphate (phosphorus) determinations
Catalog No. 50-199-5006 Supplier Hach Company Supplier No. 1436832
For accuracy checks in phosphate (phosphorus) determinations