Learn More
Hach Company Phosphate Standard Solution, 15 mgL as PO₄ (NIST), 100 mL
Catalog No. 501994968 Shop All Hach Company Products
Encompass
For accuracy checks in phosphate (phosphorus) determinations
Catalog No. 50-199-4968 Supplier Hach Company Supplier No. 1424342
For accuracy checks in phosphate (phosphorus) determinations