Learn More
Phosphate Vanadomolybdate, Reagents
Phosphate Vanadomolybdate, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$79.00 - $414.00
Chemical Identifiers
| CAS | 7732-18-5 , 7697-37-2 , 12054-85-2 |
|---|---|
| Molecular Formula | H2O |
| Molecular Weight (g/mol) | 18.02 |
| InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
| Synonym | Molybdovanadate reagent |
| IUPAC Name | water |
| SMILES | O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
01-334-523
|
Reagents Holdings Llc
CP00540010F |
10 L |
Each for $414.00
|
|
|||||
|
01-334-522
|
Reagents Holdings Llc
CP0054001A |
1 L |
Each for $79.00
|
|
|||||
Chemical Identifiers
| 7732-18-5 , 7697-37-2 , 12054-85-2 | |
| 18.02 | |
| Molybdovanadate reagent | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |
Specifications
| Yellow | |
| 100°C | |
| Molybdovanadate reagent | |
| O | |
| 18.02 | |
| Cubitainer | |
| Non-sterile | |
| Phosphate Vanadomolybdate |
| 7732-18-5 , 7697-37-2 , 12054-85-2 | |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| Reagent | |
| 1.105 | |
| Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.