Learn More
Phosphate Vanadomolybdate, Reagents
Phosphate Vanadomolybdate, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Reagents Holdings Llc CP00540010F
| Quantity | 10 L |
|---|
Chemical Identifiers
| 7732-18-5 , 7697-37-2 , 12054-85-2 | |
| 18.02 | |
| Molybdovanadate reagent | |
| O |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water |
Specifications
| Yellow | |
| 100°C | |
| Molybdovanadate reagent | |
| O | |
| 18.02 | |
| Cubitainer | |
| 10 L | |
| Liquid |
| 7732-18-5 , 7697-37-2 , 12054-85-2 | |
| H2O | |
| XLYOFNOQVPJJNP-UHFFFAOYSA-N | |
| water | |
| Reagent | |
| 1.105 | |
| Non-sterile | |
| Phosphate Vanadomolybdate |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.