CAS RN 12054-85-2
Ammonium molybdate (para) tetrahydrate, 99%
CAS: 12054-85-2 Molecular Formula: (NH4)6Mo7O24·4H2O MDL Number: MFCD00167059 Synonym: Ammonium heptamolybdate tetrahydrate; Ammonium paramolybdate tetrahydrate
Ammonium molybdate(VI) tetrahydrate, ACS reagent
CAS: 12054-85-2 Molecular Formula: H24Mo7N6O24·4H2O Molecular Weight (g/mol): 1235.86 MDL Number: MFCD00167059
Ammonium molybdate(VI) tetrahydrate, 99+%, for analysis
CAS: 12054-85-2 Molecular Formula: H24Mo7N6O24·4H2O Molecular Weight (g/mol): 1235.86
Ammonium Molybdate Tetrahydrate (Crystalline/Certified ACS), Fisher Chemical™
CAS: 12054-85-2 Molecular Formula: H24MO7N6O24 · 4 H2O MDL Number: MFCD00167059 Synonym: Ammonium Heptamolybdate Tetrahydrate
Ammonium Molybdate Tetrahydrate, ACS Reagent Grade, Ricca Chemical
CAS: 12054-85-2 Molecular Weight (g/mol): 1235.86 g/mol
Ammonium Molybdate, Tetrahydrate, MP Biomedicals™
CAS: 12054-85-2 Molecular Formula: (NH4)6Mo7O24.4H2O Synonym: Ammonium heptamolybdate tetrahydrate, Molybdic acid ammonium salt tetrahydrate
Ammonium Molybdate, Tetrahydrate, Crystal, ACS, 81.0-83.0%, Spectrum™ Chemical
CAS: 12054-85-2
Ammonium molybdate (para) tetrahydrate, ACS, 81-83% as MoO{3}
CAS: 12054-85-2 Molecular Formula: H24Mo7N6O24·4H2O MDL Number: MFCD00167059 Synonym: Ammonium paramolybdate
Ammonium molybdate tetrahydrate, For ACS analysis, 81 to 83% MoO3, MP Biomedicals™
CAS: 12054-85-2 Molecular Formula: (NH4)6 Mo7O24·4H2O Synonym: Molybdic acid ammonium salt tetrahydrate
Ammonium Molybdate, Tetrahydrate, ACS Grade, 81.0 to 83.0%, LabChem™
CAS: 12054-85-2 Molecular Weight (g/mol): 1235.86
Ammonium Molybdate RS, 5% (w/v), Ricca Chemical
CAS: 12054-85-2
Ammonium Molybdate Reagent II, Ricca Chemical
CAS: 12054-85-2 Molecular Formula: H6MoNO2 Molecular Weight (g/mol): 148.00 InChI Key: XCOMAVCHADVANQ-UHFFFAOYSA-O Synonym: Molybdic acid ammonium salt tetrahydrate,Hexammonium heptamolybdate 4-hydrate PubChem CID: 3084102 IUPAC Name: ammonium hydrate molybdenum oxidandiide SMILES: [NH4+].O.[O--].[Mo]
Ammonium Molybdate Solution, 48 g/L, Ricca Chemical
CAS: 12054-85-2 Molecular Formula: H6MoNO2 Molecular Weight (g/mol): 148.00 InChI Key: XCOMAVCHADVANQ-UHFFFAOYSA-O Synonym: Molybdic acid ammonium salt tetrahydrate,Hexammonium heptamolybdate 4-hydrate PubChem CID: 3084102 IUPAC Name: ammonium hydrate molybdenum oxidandiide SMILES: [NH4+].O.[O--].[Mo]
- 1
- 2