Learn More
CTL Scientific Supply Corp PHOSPHATESMO KM,25 PIECES
SDP
Catalog No. 501173282 Shop All CTL Scientific Supply Products
Encompass_Preferred
PHOSPHATESMO KM is a qualitative paper for the detection of sperm. In criminal investigations is used as a rapid screening test. The paper is specific for acid phosphatase. The test papers are single sealed in plastic. This ensures clear results at any time. 25 pieces (each piece 15 x 30 mm).
Catalog No. 50-117-3282 Supplier CTL Scientific Supply Corp Supplier No. 90607
PHOSPHATESMO KM is a qualitative paper for the detection of sperm. In criminal investigations is used as a rapid screening test. The paper is specific for acid phosphatase. The test papers are single sealed in plastic. This ensures clear results at any time. 25 pieces (each piece 15 x 30 mm).