Learn More
Echelon Biosciences Research Labs PHOSPHATIDIC ACID (PA) BEADS 1 mL
SDP
Supplier:  Echelon Biosciences Research Labs PB0PA1ML
Encompass
Phosphatidic Acid Beads are composed of agarose with 10 nanomoles of bound phosphatidic acid (PA) per 1ml of beads, enough for several protein binding experiments. Each 1 mL order of Phosphatidic Acid Beads includes a small amount of control beads. Larger quantities of Control Beads are also available for purchase. This item is non returnable
Catalog No. 50-822-48