Learn More
Cayman Chemical PhosphatIdylcholInbovIn 50mg
SDP
Catalog No. 502934565
Encompass_Preferred
A mixture of isolated bovine phosphatidylcholine; the most abundant phospholipids in mammalian, plant, and yeast cells; found mainly in the outer leaflet of cell membranes and can make up approximately half of the total phospholipids; a substrate for various enzymes in cell signaling pathways
Catalog No. 50-293-4565 Supplier Cayman Chemical Supplier No. 2437050 MG
A mixture of isolated bovine phosphatidylcholine; the most abundant phospholipids in mammalian, plant, and yeast cells; found mainly in the outer leaflet of cell membranes and can make up approximately half of the total phospholipids; a substrate for various enzymes in cell signaling pathways