Learn More
Cayman Chemical Phosphatidylcholine (chicken egg), Purity: ≥98%, CAS: 8002-43-5, 50 mg
SDP
Catalog No. 502932580
Encompass_Preferred
A mixture of phosphatidylcholines isolated from chicken egg; the most abundant phospholipids in mammalian, plant, and yeast cells; found mainly in the outer leaflet of cell membranes and can make up approximately half of the total phospholipids; a substrate for various enzymes in cell signaling pathways
Catalog No. 50-293-2580 Supplier Cayman Chemical Supplier No. 2434350 MG
A mixture of phosphatidylcholines isolated from chicken egg; the most abundant phospholipids in mammalian, plant, and yeast cells; found mainly in the outer leaflet of cell membranes and can make up approximately half of the total phospholipids; a substrate for various enzymes in cell signaling pathways