Learn More
Echelon Biosciences Research Labs PHOSPHATIDYLCHOLINE (PC) 100UL
SDP
Catalog No. NC1972399
Encompass
Phosphatidylcholine Beads are composed of agarose and are bound with 10 nanomoles of Phosphatidylcholine (PC) per 1 mL of beads, enough for several protein binding experiments. Each 1 mL order of Phosphatidylcholine Beads includes a small amount of Control Beads This item is non returnable
This item is not returnable. View return policy
Catalog No. NC1972399 Supplier Echelon Biosciences Research Labs Supplier No. PB0PC100UL
Phosphatidylcholine Beads are composed of agarose and are bound with 10 nanomoles of Phosphatidylcholine (PC) per 1 mL of beads, enough for several protein binding experiments. Each 1 mL order of Phosphatidylcholine Beads includes a small amount of Control Beads This item is non returnable