Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phosphatidylcholine, Spectrum™ Chemical
SDP

Catalog No. 18605163 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
10 kg
Packaging:
Amber Glass Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-163 5 g Amber Glass Bottle
18-605-164 25 g Amber Glass Bottle
18-605-165 100 g Amber Glass Bottle
18-605-166 1 kg Amber Glass Bottle
18-605-167 10 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-605-163 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P33575GM
Add to Cart
Add to Cart

P3357, 97281-47-5

Spectrum™ Chemical Phosphatidylcholine , also known as lecithin, has been shown to play a vital role in many important areas including maintaining cell structure, fat metabolism, memory, nerve signaling, as a precursor to important neurotransmitters, and liver health.

Chemical Identifiers

CAS 97281-47-5

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 5 g
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.