Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phosphatidylcholine transfer protein inhibitor-1 | 379723-99-6 | C21H17ClFN3O3S2 | 5 MG
SDP

Catalog No. 5000376025
Encompass_Preferred

Phosphatidylcholine transfer protein inhibitor-1 is a phosphatidylcholine transfer protein (PC-TP) inhibitor. It can be used for research of PC-TP related diseases such as obesity. For research use only.

  • Molecular weight: 477.96
  • Appearance: Solid
  • Color: White to off-white
  • Purity: 98.01%
  • Solubility: In DMSO, it is soluble at ≥ 100 mg/mL (209.22 mM)
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, protect from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light)

Catalog No. 50-003-76025 Supplier Medchemexpress LLC Supplier No. HY1537545MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phosphatidylcholine transfer protein inhibitor-1 is a phosphatidylcholine transfer protein (PC-TP) inhibitor. It can be used for research of PC-TP related diseases such as obesity. For research use only.

  • Molecular weight: 477.96
  • Appearance: Solid
  • Color: White to off-white
  • Purity: 98.01%
  • Solubility: In DMSO, it is soluble at ≥ 100 mg/mL (209.22 mM)
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: 4°C, protect from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.