Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Phosphatidylcholine transfer protein inhibitor-1 | 379723-99-6 | MFCD00043386 | 98.0% | 477.96 g·mol⁻¹ | C21H17ClFN3O3S2 | 10 MG
SDP

Catalog No. 5000286446
Encompass_Preferred

Phosphatidylcholine transfer protein inhibitor-1 is a small-molecule research reagent that inhibits phosphatidylcholine transfer protein (PC-TP). Supplied as a purified solid, it is intended for in vitro studies of PC-TP biology and metabolic disease research. Observe standard laboratory safety procedures when handling.

  • Selective inhibitor of phosphatidylcholine transfer protein for research applications.
  • High purity: 98.0%.
  • Molecular weight 477.96 g·mol⁻¹.
  • Chemical formula C21H17ClFN3O3S2.
  • Supplied as a 10 mg solid suitable for assay development and analytical work.
  • Intended for research use only; not for human or diagnostic use.
  • Documented with CAS 379723-99-6.

Catalog No. 50-002-86446 Supplier Medchemexpress LLC Supplier No. HY15375410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Phosphatidylcholine transfer protein inhibitor-1 is a small-molecule research reagent that inhibits phosphatidylcholine transfer protein (PC-TP). Supplied as a purified solid, it is intended for in vitro studies of PC-TP biology and metabolic disease research. Observe standard laboratory safety procedures when handling.

  • Selective inhibitor of phosphatidylcholine transfer protein for research applications.
  • High purity: 98.0%.
  • Molecular weight 477.96 g·mol⁻¹.
  • Chemical formula C21H17ClFN3O3S2.
  • Supplied as a 10 mg solid suitable for assay development and analytical work.
  • Intended for research use only; not for human or diagnostic use.
  • Documented with CAS 379723-99-6.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.