Learn More
Medchemexpress LLC Phosphatidylcholine transfer protein inhibitor-2 | 379712-59-1 | MFCD00010370 | 98.1% | 498.38 g/mol | C20H14Cl2FN3O3S2 | 10 MG
SDP
Catalog No. 5000286856
Encompass_Preferred
Phosphatidylcholine transfer protein inhibitor-2 is a small-molecule research compound that inhibits phosphatidylcholine transfer protein (PCTP). It is supplied as a purified off-white solid for biochemical and cellular studies, with reported molecular formula C20H14Cl2FN3O3S2, molecular weight 498.38 g/mol, CAS 379712-59-1, and typical purity around 98%. Store and handle according to supplier recommendations to maintain stability.
  • Inhibits phosphatidylcholine transfer protein activity in biochemical assays.
  • High reported purity supporting analytical and cell-based experiments.
  • Off-white solid amenable to dissolution in common organic solvents for assay preparation.
  • Suitable for mechanistic studies of lipid transfer and metabolic pathways.
  • Includes reported molecular and stability data to aid experimental planning.
Catalog No. 50-002-86856 Supplier Medchemexpress LLC Supplier No. HY15547310MG
Phosphatidylcholine transfer protein inhibitor-2 is a small-molecule research compound that inhibits phosphatidylcholine transfer protein (PCTP). It is supplied as a purified off-white solid for biochemical and cellular studies, with reported molecular formula C20H14Cl2FN3O3S2, molecular weight 498.38 g/mol, CAS 379712-59-1, and typical purity around 98%. Store and handle according to supplier recommendations to maintain stability.
  • Inhibits phosphatidylcholine transfer protein activity in biochemical assays.
  • High reported purity supporting analytical and cell-based experiments.
  • Off-white solid amenable to dissolution in common organic solvents for assay preparation.
  • Suitable for mechanistic studies of lipid transfer and metabolic pathways.
  • Includes reported molecular and stability data to aid experimental planning.