Learn More
Medchemexpress LLC HY-113315 50mg Medchemexpress, 3b-Hydroxy-5-cholenoic acid CAS:5255-17-4 Purity:>98%
SDP
Catalog No. 501969244
Encompass_Preferred
Medchemexpress, HY-125853 100mg Phosphatidylcholines,soya CAS:97281-47-5 Phosphatidylcholines,soya is a phosphatidylcholine from soybean used in the preparation of liposomes. Phosphatidylcholines,soya can be used as a vehicle in animal drug administration[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9244 Supplier Medchemexpress LLC Supplier No. HY11331550MG
Medchemexpress, HY-125853 100mg Phosphatidylcholines,soya CAS:97281-47-5 Phosphatidylcholines,soya is a phosphatidylcholine from soybean used in the preparation of liposomes. Phosphatidylcholines,soya can be used as a vehicle in animal drug administration[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.