Learn More
Enzo Life Sciences Phosphatidylethanol (50 mg)
SDP
Supplier:  Enzo Life Sciences BMLST4000050
Encompass_Preferred
In the presence of ethanol, phospholipase D converts phosphatidylcholine to phosphatidylethanol. This conversion is the basis of a sensitive and selective assay for PLD activity in intact cells. Phosphatidylethanol is a useful chromatographic standard for this assay. Alternative name: 1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphoethanol, PEth-16:0/18:1. Purity: ≥98% (TLC). Solubility: Soluble in chloroform. Long Term Storage: -20°C.
Catalog No. 50-201-2177