Learn More
Echelon Biosciences Research Labs PHOSPHATIDYLETHANOL BEADS 1 mL
SDP
Supplier:  Echelon Biosciences Research Labs PBPETH1ML
Encompass
Phosphatidylethanol (PEth) Beads are composed of agarose with 10 nanomoles of bound PEth per ml of beads, enough for several protein binding experiments. Each 1 mL order of PEth beads includes a small amount of control beads. Larger quantities of Control Beads are also available for purchase. This item is non returnable
Catalog No. 50-720-697