Learn More
Cayman Chemical Phosphatidylethanolamine (chicken egg), Purity: ≥98%, CAS: 39382-08-6, 50 mg
SDP
Catalog No. 502933859
Encompass_Preferred
A mixture of PEs isolated from egg; comprises approximately 25% of total mammalian phospholipids and almost 50% of brain phospholipids; found mainly in the cell membrane; synthesized mainly in the ER and mitochondrial membranes; acts as a chaperone in E. coli for lactose permease assembly; cofactor for prion propagation that can convert recombinant mammalian proteins into infectious molecules
Catalog No. 50-293-3859 Supplier Cayman Chemical Supplier No. 2433250 MG
A mixture of PEs isolated from egg; comprises approximately 25% of total mammalian phospholipids and almost 50% of brain phospholipids; found mainly in the cell membrane; synthesized mainly in the ER and mitochondrial membranes; acts as a chaperone in E. coli for lactose permease assembly; cofactor for prion propagation that can convert recombinant mammalian proteins into infectious molecules