Learn More
Cayman Chemical Phosphatidylethanolamine (soy), Purity: ≥98%, CAS: 97281-51-1, 50 mg
SDP
Catalog No. 502935528
Encompass_Preferred
A mixture of phosphatidylethanolamines isolated from soy; comprises approximately 25% of total mammalian phospholipids and almost 50% of brain phospholipids; found mainly in the cell membrane; synthesized mainly in the ER and mitochondrial membranes; acts as a chaperone in E. coli for lactose permease assembly; cofactor for prion propagation that can convert recombinant mammalian proteins into infectious molecules
Catalog No. 50-293-5528 Supplier Cayman Chemical Supplier No. 2584550 MG
A mixture of phosphatidylethanolamines isolated from soy; comprises approximately 25% of total mammalian phospholipids and almost 50% of brain phospholipids; found mainly in the cell membrane; synthesized mainly in the ER and mitochondrial membranes; acts as a chaperone in E. coli for lactose permease assembly; cofactor for prion propagation that can convert recombinant mammalian proteins into infectious molecules