Learn More
Cayman Chemical Phosphatidylglycerol (egg) (sodium salt), Purity: ≥98%, CAS: 383907-64-0, 50 mg
SDP
Catalog No. 502932759
Encompass_Preferred
A mixture of phosphatidylglycerols isolated from egg; a metabolic precursor of cardiolipin; a constituent of plant, animal, and bacterial cell membranes; species containing polyunsaturated and monounsaturated fatty acyl chains inhibit and promote proliferation of murine keratinocytes, respectively; is the second largest lipid component of mammalian lung surfactant, comprising 10% of the lipids; levels are decreased in the lung surfactant of infants with respiratory distress syndrome
Catalog No. 50-293-2759 Supplier Cayman Chemical Supplier No. 2584650 MG
A mixture of phosphatidylglycerols isolated from egg; a metabolic precursor of cardiolipin; a constituent of plant, animal, and bacterial cell membranes; species containing polyunsaturated and monounsaturated fatty acyl chains inhibit and promote proliferation of murine keratinocytes, respectively; is the second largest lipid component of mammalian lung surfactant, comprising 10% of the lipids; levels are decreased in the lung surfactant of infants with respiratory distress syndrome