Learn More
Medchemexpress LLC Phosphatidylinositols, soya, sodium salts | 383907-36-6 | 99.2% | 1 MG
SDP
Catalog No. 5000351807
Encompass_Preferred
Phosphatidylinositols, soya, sodium salts, is a mixture of phosphatidylinositols. Phosphoinositides are lipids involved in the vesicular transport of proteins and lipids between the different compartments of eukaryotic cells. They function by recruiting and/or activating effector proteins.
  • Involved in regulating various cellular functions
  • Regulates vesicular budding
  • Regulates membrane fusion
  • Regulates cytoskeleton dynamics
Catalog No. 50-003-51807 Supplier Medchemexpress LLC Supplier No. HY1395331MG
Phosphatidylinositols, soya, sodium salts, is a mixture of phosphatidylinositols. Phosphoinositides are lipids involved in the vesicular transport of proteins and lipids between the different compartments of eukaryotic cells. They function by recruiting and/or activating effector proteins.
  • Involved in regulating various cellular functions
  • Regulates vesicular budding
  • Regulates membrane fusion
  • Regulates cytoskeleton dynamics