Learn More
Medchemexpress LLC Phosphatidylinositols, soya, sodium salts | 383907-36-6 | MFCD00145109 | 99.3% | 857.0352 | C43H78NaO13P | 5 MG
SDP
Catalog No. 5000221005
Encompass_Preferred
Phosphatidylinositols, soya, sodium salts is a soy-derived mixture of phosphatidylinositol lipids provided as the sodium salt for research use. Phosphoinositides participate in membrane trafficking and lipid signaling by recruiting or activating effector proteins; this material is supplied as a solid for biochemical and lipid biology applications.
  • High reported purity (99.3%).
  • Supplied as a solid, white to off-white.
  • Available in small vial sizes suitable for assay work.
  • Suitable for studies of vesicular transport, membrane fusion, and cytoskeleton dynamics.
  • Store sealed at -20°C; in solvent, -80°C recommended for long-term storage.
Catalog No. 50-002-21005 Supplier Medchemexpress LLC Supplier No. HY1395335MG
Phosphatidylinositols, soya, sodium salts is a soy-derived mixture of phosphatidylinositol lipids provided as the sodium salt for research use. Phosphoinositides participate in membrane trafficking and lipid signaling by recruiting or activating effector proteins; this material is supplied as a solid for biochemical and lipid biology applications.
  • High reported purity (99.3%).
  • Supplied as a solid, white to off-white.
  • Available in small vial sizes suitable for assay work.
  • Suitable for studies of vesicular transport, membrane fusion, and cytoskeleton dynamics.
  • Store sealed at -20°C; in solvent, -80°C recommended for long-term storage.