Learn More
Cayman Chemical PhosphatIdylserInbovIn 100mg
SDP
Catalog No. 502935672
Encompass_Preferred
A glycerophospholipid; comprises 2-10% of total phospholipids in mammals and is enriched in the brain; found mainly on the inner leaflet of the cell membrane; synthesized in the ER and mitochondria; is a precursor to phosphatidylcholine and phosphatidylethanolamine; cofactor for various signaling pathways; externalized during apoptosis as a signal for phagocytes to engulf the cell
Catalog No. 50-293-5672 Supplier Cayman Chemical Supplier No. 24341100 MG
A glycerophospholipid; comprises 2-10% of total phospholipids in mammals and is enriched in the brain; found mainly on the inner leaflet of the cell membrane; synthesized in the ER and mitochondria; is a precursor to phosphatidylcholine and phosphatidylethanolamine; cofactor for various signaling pathways; externalized during apoptosis as a signal for phagocytes to engulf the cell