Learn More
Medchemexpress LLC Phosphatidylserine | 1446756-47-3 | 98% | C42H78NO10P | 100 MG
SDP
Catalog No. 5000432438
Encompass_Preferred
Phosphatidylserine is a well-conserved anti-inflammatory and immunosuppressive signal. It participates in Akt signaling and promotes microglial phagocytosis during synaptic refinement. It also promotes blood coagulation and acts as an "eat me" signal for apoptotic cells. This compound can suppress inflammation in tissues by inducing TGF-β secretion and inhibiting immune responses.
  • Inhibits inflammation in tissues by inducing TGF-β secretion
  • Promotes neuronal survival, differentiation, and synaptic neurotransmission
  • Mediates the pruning of microglia developmental synapses when undergoing local externalization
Catalog No. 50-004-32438 Supplier Medchemexpress LLC Supplier No. HYA0183100MG
Phosphatidylserine is a well-conserved anti-inflammatory and immunosuppressive signal. It participates in Akt signaling and promotes microglial phagocytosis during synaptic refinement. It also promotes blood coagulation and acts as an "eat me" signal for apoptotic cells. This compound can suppress inflammation in tissues by inducing TGF-β secretion and inhibiting immune responses.
  • Inhibits inflammation in tissues by inducing TGF-β secretion
  • Promotes neuronal survival, differentiation, and synaptic neurotransmission
  • Mediates the pruning of microglia developmental synapses when undergoing local externalization