Learn More
Cayman Chemical 1,2-Dipalmitoyl-sn-glycero-3-PS (sodium salt), 145849-32-7, 25 mg
SDP
Supplier:  Cayman Chemical 1508825
Encompass_Preferred
Molecular Formula C38H73NO10P • Na, Purity ≥98%, Formula Weight 758.0, Synonyms: 1,2-dipalmitoyl-sn-glycero-3-phospho-L-serine, monosodium salt, 1,2-Dipalmitoyl-sn-glycero-3-phospho-L-serine, 1,2-DPPS; A form of PS that contains the abundant long-chain (16:0) palmitic acid inserted at the sn-1 and sn-2 positions; commonly used in the generation of liposomes and other types of artificial membranes.
Catalog No. NC1003929